2-Naphthoyl chloride, 98%
2-Naphthoyl chloride, 98%
2-Naphthoyl chloride, 98%
2-Naphthoyl chloride, 98%
Acros Organics
2-Naphthoyl chloride, 98%
CAS: 2243-83-6 | C11H7ClO | 190.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number 376590500
Price (MYR)
120.00
Special offer
Online exclusive
Ends: 31-Jan-2026
169.78
Save 49.78 (29%)
50GR
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
120.00
Special offer
Online exclusive
Ends: 31-Jan-2026
169.78
Save 49.78 (29%)
50GR
Add to cart
Chemical Identifiers
CAS81-25-4
IUPAC Name(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular FormulaC24H40O5
InChI KeyBHQCQFFYRZLCQQ-OELDTZBJSA-N
SMILESC[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤103.0%
Optical Rotation≥35.0 (C=2.0 in ethanol)
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Melting Point (clear melt)197.0-204.0?C