Pentadecafluorooctanoic acid, 96%
Pentadecafluorooctanoic acid, 96%
Pentadecafluorooctanoic acid, 96%
Acros Organics
Pentadecafluorooctanoic acid, 96%
CAS: 335-67-1 | C8HF15O2 | 414.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR173961000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-50-8
IUPAC Nameiron(3+) trifluoride
Molecular FormulaF3Fe
InChI KeySHXXPRJOPFJRHA-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97% min