5-Methoxy-1-indanone, 98%
5-Methoxy-1-indanone, 98%
5-Methoxy-1-indanone, 98%
Acros Organics
5-Methoxy-1-indanone, 98%
CAS: 5111-70-6 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Catalog number ACR179670500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Methoxy-1-indanone
CAS5111-70-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0