Poly(propylene glycol), average M.W. 4.000, Thermo Scientific Chemicals
CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Have Questions?
Change view
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Catalog number ACR192200050
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(propylene glycol)
ColorGreen to Yellow
Name NoteAverage M.W. 4.000
CAS25322-69-4
Physical FormViscous Liquid