Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
Acros Organics
Dowex™ 50WX2, 200-400 mesh, ion-exchange resin
CAS: 12612-37-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic Bottle
Catalog number ACR203041000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS13734-36-6
IUPAC Name2-{[(tert-butoxy)carbonyl](methyl)amino}acetate
Molecular FormulaC8H14NO4
InChI KeyYRXIMPFOTQVOHG-UHFFFAOYSA-M
SMILESCN(CC([O-])=O)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH99.0 to 101.0 %
Water=<0.2 % (K.F.)
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals