Potassium hydrogen diiodate, 99.85%, for analysis
Potassium hydrogen diiodate, 99.85%, for analysis
Potassium hydrogen diiodate, 99.85%, for analysis
Acros Organics
Potassium hydrogen diiodate, 99.85%, for analysis
CAS: 13455-24-8 | HI2KO6 | 389.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number ACR211475000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS434-90-2
IUPAC Namedecafluoro-1,1'-biphenyl
Molecular FormulaC12F10
InChI KeyONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)65.0-72.0?C
Oxidation reagent
RUO – Research Use Only