4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
Acros Organics
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass Bottle
Catalog number ACR221130500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromophenyl isocyanate
CAS2493-02-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0