Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Actual product may vary
Fisherbrand
Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Culture AreaClosure Type
FISFB012941850 mL300 cm2Vented
LSVFB01293550 mL25 cm²Vented
FB012937250 mL75 cm²Vented
FISFB012937250 mL75 cm²Vented
LSVFB012937250 mL75 cm²Vented
FB012939600 mL182 cm2Vented
LSVFB012939600 mL182 cm2Vented
Catalog number FISFB012941
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
850 mL
Culture Area:
300 cm2
Closure Type:
Vented
  • Flat, free flask surface
  • Striation free to maximize usable growth area
  • Used in both open, closed systems
  • Angled neck design offers good pipet and cell scraper access
  • Upper triangular, wider base shape provides stability
  • Protruding ridge on back side of flask makes it easy for stacking
  • Special area near neck for easy mark writing
  • Both flask sides have engraved graduation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
  • Vented closures
  • Vented cap filter is PVDF; pore size: 0.22μm
Specifications
Certifications/ComplianceCE
Closure ColorBlue
ColorNatural
Capacity (Metric)850 mL
For Use With (Application)Cell growth
Neck StyleAngled
No. per Case18 Flasks
No. per Pack3 Flasks
Product TypeTissue Culture Flask
Surface CoatingSurface-treated
Writing SurfaceFlat
GraduatedYes
SterilitySterile
Bottom ShapeFlat
Closure TypeVented
Culture Area300 cm2
Unit Size