Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Thickness (Metric) |
|---|---|---|---|---|
| FISFB965E | 30 mL | 43mm | 37mm | 2mm |
| FKS13449699 | 30 mL | 43mm | 37mm | 2mm |
| FISFB965G | 50 mL | 53mm | 43mm | 2.1mm |
| FKS13489699 | 50 mL | 53mm | 43mm | 2.1mm |
| FKS13499699 | 100 mL | 65mm | 54mm | 2.1mm |
| FISFB965F | 40 mL | 50mm | 40mm | 2 mm |
| FKS13409709 | 40 mL | 50mm | 40mm | 2 mm |