Headspace-Vial, Crimp, Long Neck, Clear Glass
Actual product may vary
Fisherbrand
Headspace-Vial, Crimp, Long Neck, Clear Glass
Rounded or flat bottom.
Have Questions?
Catalog NumberVolume (Metric)Base Style
FKS1068084311.7 mLFlat Bottom
Catalog number FKS10680843
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
11.7 mL
Base Style:
Flat Bottom
  • Crimp Neck
  • Flat bottom
  • Long neck
  • Glass
  • Clear
Specifications
ColorClear
Diameter (Metric) Outer22.5 mm
MaterialGlass
Volume (Metric)11.7 mL
For Use With (Application)Headspace
Neck StyleCrimp
Packaging TypeCleanPack, Polypropylene Box
Volume (Metric) Residual<800 μL
Volume (Metric) Working10 mL
Base StyleFlat Bottom
Height (Metric)46 mm
Quantity100
Closure SizeND20
Marking SpotNo