Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure
Actual product may vary
Fisherbrand
Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure
Conduct trace metal analysis, high- and low-temperature work and applications involving organic solvents or rigorous cleaning with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS11311564250 mL24 mm
FIS02-923-30A125 mL24 mm
TBP1600-0004125 mL24 mm
FIS02-923-30B250 mL24 mm
Catalog number FKS11311564
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Size:
24 mm
Conduct trace metal analysis, high- and low-temperature work and applications involving organic solvents or rigorous cleaning with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure.
  • Autoclavable and transparent
  • Chemical and corrosion resistant
  • Withstand -105°C to +150°C (-157°F to +302°F)
  • Leakproof†, ETFE screw caps included
  • Completely disengage threads or remove cap before autoclaving
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
DescriptionNarrow Mouth Bottle
Diameter (Metric) Inner Neck17 mm
For Use With (Application)Storage, Transportation ​
MaterialTeflon FEP
No. per Case4/Cs.
No. per Pack1 Ea.
SterilityNon-sterile
Temperature Range (Metric)-100°C to 260°C
Closure ColorNatural
Neck Finish24 mm
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)134 mm
Product TypeNarrow Mouth Bottle
Width (English)2.36 in.
Height (English)5.28 in.
Closure MaterialETFE
Closure Size24 mm
Diameter (Metric)60 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
AutoclavableAutoclavable