Cimarec+™ Stirring Hotplates Series
Actual product may vary
Fisherbrand
Cimarec+™ Stirring Hotplates Series
Get precise stirring control, exceptional safety and temperature performance for your routine protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTop Plate MaterialSizeElectrical RequirementsPlug Type
FKS15483600Ceramic4.25 x 4.25 in.230 V 50/60 HzEU/UK/AUS/CN
SP88854105Ceramic4.25 x 4.25 in.230 V 50/60 HzEU/UK/AUS/CN
SP88857105Ceramic7.25 x 7.25 in.230 V 50/60 HzEU
SP88857107Ceramic7.25 x 7.25 in.230 V 50/60 HzUK
SP88857108Aluminum7.25 x 7.25 in.230 V 50/60 HzEU/UK/AUS/CN
SP88850105Ceramic10.25 x 10.25 in.230 V 50/60 HzEU/UK
Catalog number FKS15483600
Price (MYR)
7,544.00
Each
Top Plate Material:
Ceramic
Size:
4.25 x 4.25 in.
Electrical Requirements:
230 V 50/60 Hz
Plug Type:
EU/UK/AUS/CN
Price (MYR)
7,544.00
Each
Get precise stirring control for your routine protocols using Thermo Scientific™ Cimarec+™ series digital stirrers. The Cimarec+ Stirring Hotplate Series are available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring, besides 4 x 4 in. Hotplate Stirrer, which has digital temperature and analog stirring control.
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Warranty: 3 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Top Plate MaterialCeramic
Voltage230 V
Includes(3) Power Cords
Shipping Weight (English)8.4 lb.
Shipping Weight (Metric)3.8 kg
Electrical Requirements230 V 50/60 Hz
Length (English)13 in.
Size4.25 x 4.25 in.
Height (Metric)9.7 cm
Temperature (Metric)Ambient to 540°C
Height (English)3.8 in.
Frequency50/60 Hz
Width (Metric)20.8 cm
Length (Metric)33 cm
Width (English)8.2 in.
Weight (English)11.2 lb.
Weight (Metric)5.1 kg
TypeStirring Hotplate
ShapeSquare
Plug TypeEU/UK/AUS/CN
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (L x W x H)10 x 5 x 3.6 in. (25.4 x 12.7 x 9.1 cm)
Certifications/CompliancecCSAus, CE
DisplayLED
Unit SizeEach