3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
Alfa Aesar
3-Amino-1H-indazole, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA435740010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Amino-1H-indazole
CAS874-05-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0