Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
Alfa Aesar
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA437200010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1317-36-8
IUPAC Nameplumbanone
Molecular FormulaOPb
InChI KeyYEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILESO=[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light orange
Average particle size=<10 µm
Total trace metal impurities=<1000.0 ppm
Appearance (Form)Powder
For ultra trace analysis
RUO – Research Use Only