Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals
Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals
Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals
Alfa Aesar
Benzyl alcohol, specified according to requirements of Ph.Eur., Thermo Scientific Chemicals
CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA447000010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS959-36-4
IUPAC Name(6E)-6-[(2-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}hydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Molecular FormulaC14H12N2O2
InChI KeySPEXYYIULCBQJR-WGDLNXRISA-N
SMILESO=C1C=CC=C\C1=C/NN\C=C1/C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Melting Point (clear melt)214.0-224.0?C
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Appearance (Color)Yellow