Zinc acetate dihydrate, 97%, specified according to the requirements of USP, Thermo Scientific Chemicals
Zinc acetate dihydrate, 97%, specified according to the requirements of USP, Thermo Scientific Chemicals
Zinc acetate dihydrate, 97%, specified according to the requirements of USP, Thermo Scientific Chemicals
Alfa Aesar
Zinc acetate dihydrate, 97%, specified according to the requirements of USP, Thermo Scientific Chemicals
CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA451880010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc acetate dihydrate
Melting Point237.0°C
CAS5970-45-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Very toxic to aquatic life.