D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
Alfa Aesar
D(-)-Fructose, specified according the requirements of USP, Thermo Scientific Chemicals
CAS: 57-48-7 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number FSA451962500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to light yellow
Appearance (Form)Solution
Titration with HClO4>=40 %