Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
Alfa Aesar
Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
CAS: 7758-09-0 | KNO2 | 85.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA453190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS1757-42-2
IUPAC Name3-methylcyclopentan-1-one
Molecular FormulaC6H10O
InChI KeyAOKRXIIIYJGNNU-UHFFFAOYNA-N
SMILESCC1CCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4325-1.4355 @ 20?C