Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
Alfa Aesar
Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
CAS: 7550-35-8 | BrLi | 86.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic Bottle
Catalog number FSA453980010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Boiling Point1265.0°C
Chemical Name or MaterialLithium Bromide
Melting Point550.0°C
CAS7550-35-8
Health Hazard 1GHS Signal Word: Warning
View more
Suitable for battery materials development.
RUO – Research Use Only