Specifications
Certifications/ComplianceASTM D5151, ASTM D412, ASTM D573, ASTM D3767, ASTM D3577, ASTM D3767, ASTM D6319, IEST-RP-CC005, ASTM D6978-05
Certifications/Compliance (Europe)EN ISO 374-1:2016; EN 374-4:2003; EN ISO 374-5:2016
Chemotherapeutic RatedYes
Cleanroom ClassClass 10 (ISO 4)
Accelerator FreeYes
AQL1.5
ColorWhite
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
HandSpecific
Latex FreeYes
MaterialNitrile
Palm Thickness (English)5.1 mil
Palm Thickness (Metric)0.13 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size6.5
SterilitySterile
Thickness (English)6 mil
Thickness (Metric)0.15 mm
Model56889
SeriesKimtech™ G3 Sterile
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase