ColorWhite
MaterialCardboard
ShapeSquare
TypeCryo Box
For Use With (Application)Cryogenic Storage of Liquids
Includes9 x 9 Divider
Quantity36 Pack
Dimensions (L x W x H)133 x 133 x 52 mm
Length (Metric)133 mm
Width (Metric)133 mm
Height (Metric)52 mm
Weight (Metric)3.38 kg
Unit SizePack